Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Rab20 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Rab20 Polyclonal specifically detects Rab20 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Rab20 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
FLJ20429, RAB20, member RAS oncogene family, ras-related protein Rab-20 | |
RAB20 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
55647 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLQRYMERRFPDTVSTVGGAFYLKQWRSYNISIWDTAGREQFHGLGSMYCR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title