Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab20 Antibody (OTI1F1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273768G
Description
Rab20 Monoclonal specifically detects Rab20 in Human samples. It is validated for Western Blot.Specifications
Rab20 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB20 | |
Full length recombinant protein of human Rab20 (NP_060287) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI1F1 | |
Western Blot | |
FLJ20429, RAB20, member RAS oncogene family, ras-related protein Rab-20 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
55647.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction