Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551042
Description
RAB20 Polyclonal Antibody detects endogenous levels of RAB20 protein.
Plays a role in apical endocytosis/recycling. Plays a role in the maturation and acidification of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis. Plays a role in the fusion of phagosomes with lysosomes. [UniProt].
Specifications
RAB20 | |
Polyclonal | |
Unconjugated | |
Rab20 | |
AA536966; D8Ertd350e; RAB20; RAB20, member RAS oncogene family; ras-related protein Rab-20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55647 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NX57 | |
Rab20 | |
Synthesized peptide derived from the Internal region of human Rab 20 (aa 115-164). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction