Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB21 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282030MFV610
Description
RAB21 Polyclonal antibody specifically detects RAB21 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| RAB21 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| KIAA0118ras-related protein Rab-21, RAB21, member RAS oncogene family | |
| RAB21 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human RAB21. The immunogen is located within the last 50 amino acids of RAB21. Amino Acid Squence: QVDERAKGNGSSQPGTARR | |
| 0.1 mL | |
| Signal Transduction | |
| 23011 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction