Learn More
Invitrogen™ RAB21 Monoclonal Antibody (OTI6E12)

Description
RAB21 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | RAB21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI6E12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RAB21 |
| Gene Accession No. | P35282, P55745, Q6AXT5, Q9UL25 |
| Gene Alias | 9630024B22; GTP-binding protein RAB21; KIAA0118; Rab-12; Rab21; RAB21, member RAS oncogene family; ras-related protein Rab-21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.