Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB22A Antibody (SR1340), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32263350UL
Description
RAB22A Monoclonal antibody specifically detects RAB22A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation, SerumSpecifications
RAB22A | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
SR1340 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Serum 1:50 | |
GTP-binding protein RAB22A, MGC16770, Rab-22, RAB22, RAB22A, member RAS oncogene family, ras-related protein Rab-22A | |
A synthesized peptide derived from human RAB22A (Uniprot #: Q9UL26) | |
50 μg | |
Signal Transduction | |
57403 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction