Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RAB23 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RAB23 Polyclonal specifically detects RAB23 in Human samples. It is validated for Western Blot.Specifications
| RAB23 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23 | |
| RAB23 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9ULC3 | |
| 51715 | |
| Synthetic peptides corresponding to RAB23(RAB23, member RAS oncogene family) The peptide sequence was selected from the N terminal of RAB23. Peptide sequence TKDYKKTIGVDFLERQIQVNDEDVRLMLWDTAGQEEFDAITKAYYRGAQA. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title