Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab24 Antibody (OTI3H1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273770G
Description
Rab24 Monoclonal specifically detects Rab24 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Rab24 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB24 | |
Full length recombinant protein of human Rab24 (NP_001026847) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Canine | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI3H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
RAB24, member RAS oncogene family, ras-related protein Rab-24 | |
Mouse | |
Affinity Purified | |
Autophagy, Signal Transduction | |
53917.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction