Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB24 Monoclonal Antibody (OTI3H1)
GREENER_CHOICE

Catalog No. PIMA526188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526188 Supplier Invitrogen™ Supplier No. MA526188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB24 Monoclonal Antibody for Western Blot, ICC/IF

RAB24 may be involved in autophagy-related processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB24
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB24
Gene Accession No. P35290, Q969Q5
Gene Alias 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24
Gene Symbols RAB24
Host Species Mouse
Immunogen Full length human recombinant protein of RAB24 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19336, 479277, 53917
Target Species Human, Mouse, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.