Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145189 Supplier Invitrogen™ Supplier No. PA5145189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAAIVCYDLT DSSSFERAKF WVKELRSLEE GCQIYLCGTK SDLLEEDRRR RRVDFHDVQD YA Antigen sequence identity: Mouse 100%; Rat 100%.

RAB24 may be involved in autophagy-related processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB24
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB24
Gene Accession No. Q969Q5
Gene Alias 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24
Gene Symbols RAB24
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 82-143. Recombinant protein control fragment (Product #RP-109794).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53917
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.