Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117130 Supplier Invitrogen™ Supplier No. PA5117130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-1080, MCF7, SH-SY5Y, 293T, Mouse brain Immunogen sequence: MSGQRVDVKV VMLGKEYVGK TSLVERYVHD RFLVGPYQNT IGAAFVAKVM SVGDRTVTLG IWDTAGSERY EAMSRIYYRG AKAAIVCYDL TDSSSFERAK FWVKELRSLE EGCQIYLCGT KSDLLEEDRR RRRVDFHDVQ DYADNIKAQL FETSSKTGQS VDELFQKVAE DYVSVAAFQV MTEDKGVDLG QKPNPYFYSC CHH.

RAB24 may be involved in autophagy-related processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB24
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB24
Gene Accession No. P35290, Q969Q5
Gene Alias 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24
Gene Symbols RAB24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-203 of human RAB24 (NP_0010268471).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19336, 361208, 53917
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.