Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab25 Antibody (SR1367), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32195450UL
Description
Rab25 Monoclonal antibody specifically detects Rab25 in Human samples. It is validated for Western BlotSpecifications
Rab25 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
57111 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR1367 | |
Western Blot 1:500-1:1000 | |
CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
A synthesized peptide derived from human Rab25 (Uniprot #: P57735) | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction