Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB25 Monoclonal Antibody (3F12)
GREENER_CHOICE

Catalog No. PIMA515587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515587 Supplier Invitrogen™ Supplier No. MA515587
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15587 targets Rab25 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human and mouse samples. The MA5-15587 immunogen is purified recombinant fragment of Rab25 expressed in E. Coli. MA5-15587 detects Rab25 which has a predicted molecular weight of approximately 24kDa.

RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB25
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3F12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene RAB25
Gene Accession No. P57735, Q9WTL2
Gene Alias AV012978; CATX8; CATX-8; RAB11C; RAB25; RAB25, member RAS oncogene family; ras-related protein Rab-25
Gene Symbols RAB25
Host Species Mouse
Immunogen Purified recombinant fragment of Rab25 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53868, 57111
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.