Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB25 Monoclonal Antibody (3F12F3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515267
Description
MA5-15267 targets Rab25 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15267 immunogen is purified recombinant fragment of Rab25 expressed in E. Coli.
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
Specifications
RAB25 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P57735 | |
RAB25 | |
Purified recombinant fragment of Rab25 expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3F12F3 | |
Unconjugated | |
RAB25 | |
AV012978; CATX8; CATX-8; RAB11C; RAB25; RAB25, member RAS oncogene family; ras-related protein Rab-25 | |
Mouse | |
Protein G | |
RUO | |
57111 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction