Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110256 Supplier Invitrogen™ Supplier No. PA5110256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGNGTEEDYN FVFKVVLIGE SGVGKTNLLS RFTRNEFSHD SRTTIGVEFS TRTVMLGTAA VKAQIWDTAG LERYRAITSA YYRGAVGALL VFDLTKHQTY AVVERWLKEL YDHAEATIVV MLVGNKSDLS QAREVPTEEA RMFAENNGLL FLETSALDST NVELAFETVL KEIFAKVSKQ RQNSIRTNAI TLGSAQAGQE PGPGEKRACC ISL.

RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB25
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAB25
Gene Accession No. Q9WTL2
Gene Alias AV012978; CATX8; CATX-8; RAB11C; RAB25; RAB25, member RAS oncogene family; ras-related protein Rab-25
Gene Symbols RAB25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-213 of human RAB25 (NP_065120.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310632, 53868
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.