Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544261
Description
Peptide sequence: SRKKTPKSKG ASTPAASTLP TANGARPARS GTALSGPDAP PNGPLQPGRP Sequence homology: Human: 100%; Rat: 79%.
Members of the RAB protein family, including RAB26, are important regulators of vesicular fusion and trafficking. The RAB family of small G proteins regulates intercellular vesicle trafficking, including exocytosis, endocytosis, and recycling (summary by Seki et al., 2000 ).
Specifications
RAB26 | |
Polyclonal | |
Unconjugated | |
RAB26 | |
A830020M03Rik; RAB26; RAB26, member RAS oncogene family; Ras-related oncogene protein; ras-related protein Rab-26; V46133 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25837 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9ULW5 | |
RAB26 | |
Synthetic peptide directed towards the N-terminal of human RAB26 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction