Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116683 Supplier Invitrogen™ Supplier No. PA5116683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, Mouse pancreas, Rat kidney Immunogen sequence: MSRKKTPKSK GASTPAASTL PTANGARPAR SGTALSGPDA PPNGPLQPGR PSLGGGVDFY.

Members of the RAB protein family, including RAB26, are important regulators of vesicular fusion and trafficking. The RAB family of small G proteins regulates intercellular vesicle trafficking, including exocytosis, endocytosis, and recycling (summary by Seki et al., 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB26
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAB26
Gene Accession No. P51156, Q504M8, Q9ULW5
Gene Alias A830020M03Rik; RAB26; RAB26, member RAS oncogene family; Ras-related oncogene protein; ras-related protein Rab-26; V46133
Gene Symbols RAB26
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-60 of human RAB26 (NP_0551682).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171111, 25837, 328778
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.