Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab27a Antibody (2E8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Rab27a |
---|---|
Clone | 2E8 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Rab27a Monoclonal antibody specifically detects Rab27a in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Rab27a | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A | |
A synthesized peptide derived from Human Rab27a [UniProt P51159] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2E8 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5873 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title