Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB27A Monoclonal Antibody (1590CT813.266.26)
GREENER_CHOICE

Catalog No. PIMA537692
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537692 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537692 Supplier Invitrogen™ Supplier No. MA537692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB27A Monoclonal Antibody for Flow, Western Blot

The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB27A
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1590CT813.266.26
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB27A
Gene Accession No. P51159
Gene Alias 2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; ashen; GS2; GTP-binding protein Ram; HsT18676; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; RAB27; rab-27; Rab27a; RAB27A, member RAS oncogene family; ram; Ram p25; ras-related protein Rab-27A
Gene Symbols RAB27A
Host Species Mouse
Immunogen recombinant protein of human RAB27A.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.