Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB27A Mouse anti-Human, Unconjugated, Clone: 1G7, Abnova™

Catalog No. 89335109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-109 Supplier Abnova Corporation Supplier No. H00005873M02J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

RAB27A Monoclonal antibody specifically detects RAB27A in Human samples. It is validated for WB-Ce, IHC-P, S-ELISA, ELISA, WB-Re, WB-Tr, RNAi-Ab.

Specifications

Antigen RAB27A
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA, KnockDown
Classification Monoclonal
Clone 1G7
Conjugate Unconjugated
Description RAB27A, member RAS oncogene family
Gene RAB27A
Gene Accession No. NM_004580
Gene Alias GS2, HsT18676, MGC117246, RAB27, RAM
Host Species Mouse
Immunogen RAB27A (NP_004571, 122 a.a. ∼ 221 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5873
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.