Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB27A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544580 Supplier Invitrogen™ Supplier No. PA544580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SQAIEMLLDL IMKRMERCVD KSWIPEGVVR SNGHASTDQL SEEKEKGACG Sequence homology: Cow: 79%; Goat: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Rat: 93%; Sheep: 79%.

The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB27A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAB27A
Gene Accession No. P51159
Gene Alias 2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; ashen; GS2; GTP-binding protein Ram; HsT18676; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; RAB27; rab-27; Rab27a; RAB27A, member RAS oncogene family; ram; Ram p25; ras-related protein Rab-27A
Gene Symbols RAB27A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RAB27A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5873
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.