Learn More
Invitrogen™ RAB27A Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547907
Description
This antibody detects human, mouse, and rat RAB27A in Western blots and recombinant human RAB27A in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human RAB27B is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
Specifications
RAB27A | |
Polyclonal | |
Unconjugated | |
RAB27A | |
2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; ashen; GS2; GTP-binding protein Ram; HsT18676; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; RAB27; rab-27; Rab27a; RAB27A, member RAS oncogene family; ram; Ram p25; ras-related protein Rab-27A | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
11891, 50645, 5873 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P23640, P51159, Q9ERI2 | |
RAB27A | |
E. coli-derived recombinant human RAB27A Ser135-Ala218. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.