Learn More
Invitrogen™ RAB27B Monoclonal Antibody (1596CT245.254.49)

Description
RAB27B Monoclonal Antibody for Flow, Western Blot
Specifications
Specifications
| Antigen | RAB27B |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 1596CT245.254.49 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RAB27B |
| Gene Accession No. | O00194, Q99P58 |
| Gene Alias | 2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.