Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB27B Monoclonal Antibody (1596CT584.1.45)
GREENER_CHOICE

Catalog No. PIMA524743
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524743 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524743 Supplier Invitrogen™ Supplier No. MA524743
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB27B Monoclonal Antibody for Western Blot

RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB27B
Applications Western Blot
Classification Monoclonal
Clone 1596CT584.1.45
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB27B
Gene Accession No. O00194, Q99P58
Gene Alias 2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1
Gene Symbols RAB27B
Host Species Mouse
Immunogen human RAB27B recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5874, 80718
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.