Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB27B Mouse anti-Human, Clone: 4G7, Bioss
SDP

Catalog No. 501979848
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9848 Supplier Bioss Supplier No. BSM51331M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB27B may be involved in targeting uroplakins to urothelial apical membranes.

Specifications

Antigen RAB27B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4G7
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene RAB27B
Gene Accession No. O00194
Gene Alias 2310021G14Rik; B130064M09Rik; BOS_22239; C25KG; LOC100343056; Rab27b; RAB27B, member RAS oncogene family; Ras-related protein Rab-27B
Gene Symbols RAB27B
Host Species Mouse
Immunogen This RAB27B antibody is generated from a mouse immunized with a recombinant protein of human RAB27B.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5874
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.