Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB27B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524908
Description
RAB27B Polyclonal Antibody for Western Blot, Flow
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
Specifications
RAB27B | |
Polyclonal | |
Unconjugated | |
RAB27B | |
2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O00194 | |
RAB27B | |
KLH conjugated synthetic peptide between 186-215 amino acids from the C-terminal region of human RAB27B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction