Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB27B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570223 Supplier Invitrogen™ Supplier No. PA570223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB27B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAB27B
Gene Accession No. O00194, Q99P58
Gene Alias 2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1
Gene Symbols RAB27B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RAB27B (aa 122-171).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5874, 80718
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.