Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB27B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119699
This item is not returnable.
View return policy
Description
Positive Control: U937 cell lysates, SH-SY5Y. Subcellular Location: Late endosome, Membrane, Lipid-anchor.
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
Specifications
| RAB27B | |
| Polyclonal | |
| Unconjugated | |
| RAB27B | |
| 2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O00194 | |
| RAB27B | |
| Synthetic peptide with N-terminal Human RAB27B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction