Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548813
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548813 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548813 Supplier Invitrogen™ Supplier No. PA548813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB28 Polyclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the Rab subfamily of Ras-related small GTPases. The encoded protein may be involved in regulating intracellular trafficking. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 9 and X. [provided by RefSeq, Apr 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB28
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB28
Gene Accession No. P51157
Gene Alias 2700023P08Rik; AW496496; CORD18; rab-26; Rab28; RAB28, member RAS oncogene family; ras-related protein Rab-28
Gene Symbols RAB28
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 119-147 amino acids from the Central region of human RAB28.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9364
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.