Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104071
Description
Antibody detects endogenous levels of total RAB28.
This gene encodes a member of the Rab subfamily of Ras-related small GTPases. The encoded protein may be involved in regulating intracellular trafficking. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 9 and X. [provided by RefSeq, Apr 2009].
Specifications
RAB28 | |
Polyclonal | |
Unconjugated | |
RAB28 | |
2700023P08Rik; AW496496; CORD18; rab-26; Rab28; RAB28, member RAS oncogene family; ras-related protein Rab-28 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100972, 117049, 9364 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P51157, P51158, Q99KL7 | |
RAB28 | |
A synthesized peptide derived from human RAB28(Accession P51157), corresponding to amino acid residues T109-S159. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction