Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531007 Supplier Invitrogen™ Supplier No. PA531007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Raji. Predicted reactivity: Mouse (91%), Rat (93%), Rhesus Monkey (97%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Rab GTPase key regulator in vesicle trafficking. Essential for maintaining the integrity of the endosome-trans-Golgi network structure. Together with LRRK2, plays a role in the retrograde trafficking pathway for recycling proteins, such as mannose 6 phosphate receptor (M6PR), between lysosomes and the Golgi apparatus in a retromer-dependent manner. Regulates neuronal process morphology in the intact central nervous system (CNS). May play a role in the formation of typhoid toxin transport intermediates during Salmonella enterica serovar Typhi (S.Typhi) epithelial cell infection. {ECO:0000269; PubMed:22042847, ECO:0000269; PubMed:24788816}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB29
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RAB29
Gene Accession No. O14966
Gene Alias RAB29; RAB29, member RAS oncogene family; RAB7, member RAS oncogene family-like 1; RAB7L; RAB7L1; Rab-7-like protein 1; Ras-related GTP-binding protein Rab29; ras-related protein Rab-29; Ras-related protein Rab-7L1
Gene Symbols RAB29
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB7L1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8934
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.