Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588190 Supplier Invitrogen™ Supplier No. PA588190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAYAYLFKYI IIGDTGVGKS CLLLQFTDKR FQPVHDLTIG VEFGARMITI DGKQIKLQIW DTAGQESFRS ITRSYYRGAA GALLVYDITR RDTFNHLTTW LEDARQHSNS NMVIMLIGNK SDLESRREVK KEEGEAFARE HGLIFMETSA KTASNVEEAF INTAKEIYEK IQEGVFDINN EANGIKIGPQ HAATNATHAG NQGGQQAGGG CC; Positive Samples: U-87MG, HeLa, HT-29, A-549, Mouse lung, Mouse testis, Rat brian; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Golgi apparatus membrane, Lipid-anchor, Melanosome.

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB2A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Rab2a
Gene Accession No. P05712, P53994, P61019
Gene Alias 9330148M11Rik; C80220; GTP binding protein; GTP-binding protein; LHX; OTTMUSP00000004796; rab 2; RAB2; RAB2, member RAS oncogene family; Rab2a; RAB2A, member RAS oncogene family; ras-related protein Rab-2A; RP23-183L6.1; small GTP binding protein RAB2A
Gene Symbols Rab2a
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5862, 59021, 65158
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.