Learn More
RAB2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA530142
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (100%), Xenopus laevis (88%), Dog (100%), Rabbit (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.
Specifications
RAB2A | |
Polyclonal | |
Unconjugated | |
Rab2a | |
9330148M11Rik; C80220; GTP binding protein; GTP-binding protein; LHX; OTTMUSP00000004796; rab 2; RAB2; RAB2, member RAS oncogene family; Rab2a; RAB2A, member RAS oncogene family; ras-related protein Rab-2A; RP23-183L6.1; small GTP binding protein RAB2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.42 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P61019 | |
Rab2a | |
Recombinant fragment corresponding to a region within amino acids 1 and 177 of Human RAB2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.