Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB2B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA562165

Catalog No. PIPA562165


Only null left
Add to Cart

Description

Description

Immunogen sequence: GIKIGPQQSI STSVGPSASQ RNSRDIGSNS GCC Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 76%.

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAB2B
Polyclonal
Unconjugated
RAB2B
1500012D09Rik; 4930528G15Rik; A230002G14; D530043M21Rik; GTP-binding protein RAB2B; Rab2b; RAB2B, member RAS oncogene family; RAS family, member RAB2B; ras-related protein Rab-2B
Rabbit
Antigen affinity chromatography
RUO
84932
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WUD1
RAB2B
Recombinant protein corresponding to Human RAB2B. Recombinant protein control fragment (Product #RP-101282).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.