Learn More
Rab3-GAP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302114AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Rab 3 GAP p130, also known as Rab3 GTPase-activating protein catalytic subunit, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.Specifications
Rab3-GAP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15042 | |
RAB3GAP1 | |
Between 575 and 625 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RAB3GAP1 | |
Rab3 GTPase-activating protein catalytic subunit, RAB3 GTPase-activating protein 130 kDa subunit, Rab3-GAP p130, Rab3-GAP, KIAA0066, RAB3GAP, Rab 3 GAP p130, Rab3 GAP p130 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22930 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.