Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB30 Antibody (OTI3E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245498
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
RAB30 Monoclonal specifically detects RAB30 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RAB30 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
RAB30, member RAS oncogene family, ras-related protein Rab-30 | |
Mouse | |
22.9 kDa | |
0.1 mL | |
Signal Transduction | |
27314 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI3E7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RAB30 | |
Full length human recombinant protein of human RAB30(NP_055303) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction