Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB30 Monoclonal Antibody (OTI3E7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526112
Description
RAB30 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.
Specifications
RAB30 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15771, Q923S9 | |
RAB30 | |
Full length human recombinant protein of RAB30 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI3E7 | |
Unconjugated | |
RAB30 | |
5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27314, 308821, 476787, 75985 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction