Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB30 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505343S


Description
RAB30 Monoclonal antibody specifically detects RAB30 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
RAB30 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15771 | |
Mouse | |
Affinity chromatography | |
RUO | |
27314 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4B9 | |
Unconjugated | |
RAB30 | |
5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30 | |
Full length human recombinant protein of human RAB30 produced in HEK239T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction