Learn More
RAB30 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3F4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505341

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.Specifications
RAB30 | |
Monoclonal | |
Unconjugated | |
RAB30 | |
RAB30, member RAS oncogene family | |
Mouse | |
Affinity Chromatography | |
RUO | |
27314, 308821, 476787, 75985 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3F4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15771, Q923S9 | |
Rab30 | |
Full length human recombit protein of human RAB30 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.