Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537206
Description
This antibody detects endogenous protein at a molecular weight of 23 kDa. Purity is >95% by SDS-PAGE.
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.
Specifications
| RAB30 | |
| Polyclonal | |
| Unconjugated | |
| RAB30 | |
| 5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27314, 308821, 75985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
| Q15771, Q923S9 | |
| RAB30 | |
| A synthetic peptide corresponding to residues in human RAB30. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction