Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB31 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297073PE
Description
RAB31 Polyclonal antibody specifically detects RAB31 in Human samples. It is validated for ELISASpecifications
RAB31 | |
Polyclonal | |
ELISA | |
Rab22B, RAB31, member RAS oncogene family, Ras-related protein Rab-22B, ras-related protein Rab-31 | |
Rabbits immunized with purified, recombinant Human RAB31 (Accession#: NP_006859.2, Met1-Cys195) | |
0.1 mL | |
Signal Transduction | |
11031 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction