Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554064
Description
Immunogen sequence: LKDAKEYAES IGAIVVETSA KNAINIEELF QGISRQIPPL DPHENGNNGT IKVEKPTMQA SRRCC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].
Specifications
RAB31 | |
Polyclonal | |
Unconjugated | |
RAB31 | |
1700093E07Rik; AI415285; GTP-binding protein Rab0; monomeric GTP-binding protein; Rab0; Rab22B; Rab31; RAB31, member RAS oncogene family; ras-related protein Rab-22B; Ras-related protein Rab-31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
106572, 11031, 246324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13636, Q6GQP4, Q921E2 | |
RAB31 | |
Recombinant protein corresponding to Human RAB31. Recombinant protein control fragment (Product #RP-91757). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction