Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110258 Supplier Invitrogen™ Supplier No. PA5110258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMAIRELKVC LLGDTGVGKS SIVCRFVQDH FDHNISPTIG ASFMTKTVPC GNELHKFLIW DTAGQERFHS LAPMYYRGSA AAVIVYDITK QDSFYTLKKW VKELKEHGPE NIVMAIAGNK CDLSDIREVP LKDAKEYAES IGAIVVETSA KNAINIEELF QGISRQIPPL DPHENGNNGT IKVEKPTMQA SRRCC.

Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB31
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB31
Gene Accession No. Q13636, Q6GQP4, Q921E2
Gene Alias 1700093E07Rik; AI415285; GTP-binding protein Rab0; monomeric GTP-binding protein; Rab0; Rab22B; Rab31; RAB31, member RAS oncogene family; ras-related protein Rab-22B; Ras-related protein Rab-31
Gene Symbols RAB31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human RAB31 (NP_006859.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106572, 11031, 246324
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.