Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548865
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548865 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548865 Supplier Invitrogen™ Supplier No. PA548865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB32 Polyclonal Antibody for Western Blot

RAB32 acts as an A-kinase anchoring protein by binding to the type II regulatory subunit of protein kinase A and anchoring it to the mitochondrion. Also involved in synchronization of mitochondrial fission. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB32
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB32
Gene Accession No. Q13637
Gene Alias 2810011A17Rik; AU022057; RAB32; RAB32, member RAS oncogene family; ras-related protein Rab-32; RGD1559997
Gene Symbols RAB32
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 2-28 amino acids from the N-terminal region of human RAB32.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.