Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB33A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101826
Description
Antibody detects endogenous levels of total RAB33A.
The Ras-related superfamily of guanine nucleotide binding proteins includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies. Increasing data suggests an important role for Rab proteins in either endocytosis or in biosynthetic protein transport. The process of transporting newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves the movement of carrier vesicles and requires Rab protein function. Rab proteins are also an integral part of endocytic pathways. Rab 33B and Rab 33A share conserved effector domains, but share only 55.3% overall amino acid identity. This suggests that Rab 33B and Rab 33A may interact with similar effector and regulatory proteins.
Specifications
RAB33A | |
Polyclonal | |
Unconjugated | |
RAB33A | |
RAB 33A; Rab33a; RAB33A, member of RAS oncogene family; RAB33A, member RAS oncogene family; Rabs10; Ras-related protein Rab-33A; RGD1563280; small GTP-binding protein S10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
19337, 317580, 9363 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97950, Q14088 | |
RAB33A | |
A synthesized peptide derived from human RAB33A(Accession Q14088), corresponding to amino acid residues I132-A182. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction