Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB33B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530697 Supplier Invitrogen™ Supplier No. PA530697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, MCF-7, MDA-MB-231, Mouse brain. Predicted reactivity: Mouse (96%), Rat (97%), Xenopus laevis (80%), Chicken (94%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Small GTP-binding proteins of the RAB family, such as RAB33B, play important roles at defined steps of vesicular transport in protein secretion and the endocytosis pathway (Zheng et al., 1998 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB33B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RAB33B
Gene Accession No. O35963, Q9H082
Gene Alias RAB33B; RAB33B, member of RAS oncogene family; RAB33B, member RAS oncogene family; ras-related protein Rab-33B; ras-related protein Rab-33B; LOW QUALITY PROTEIN: ras-related protein Rab-33B; SMC2
Gene Symbols RAB33B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB33B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100685754, 19338, 365793, 83452
Target Species Canine, Human, Mouse, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.