Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB33B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104074
Description
Antibody detects endogenous levels of total RAB33B.
Small GTP-binding proteins of the RAB family, such as RAB33B, play important roles at defined steps of vesicular transport in protein secretion and the endocytosis pathway (Zheng et al., 1998 ).
Specifications
RAB33B | |
Polyclonal | |
Unconjugated | |
RAB33B | |
RAB33B; RAB33B, member of RAS oncogene family; RAB33B, member RAS oncogene family; ras-related protein Rab-33B; ras-related protein Rab-33B; LOW QUALITY PROTEIN: ras-related protein Rab-33B; SMC2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19338, 365793, 83452 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35963, Q9H082 | |
RAB33B | |
A synthesized peptide derived from human RAB33B(Accession Q9H082), corresponding to amino acid residues I129-A179. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction