Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599697 Supplier Invitrogen™ Supplier No. PA599697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Rab 34.

Protein transport. Involved in the redistribution of lysosomes to the peri-Golgi region. Plays a role in the maturation of phagosomes that engulf pathogens, such as S. aureus and M. tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB34
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB34
Gene Accession No. Q5U1Y1, Q9BZG1
Gene Alias AI326479; GTP-binding protein rah; Narr; Nine amino-acid residue-repeats; RAB34; RAB34, member of RAS oncogene family; RAB34, member RAS oncogene family; RAB39; rab-39; RAH; Rah1; Ras-related homolog; Ras-related protein RAB34; ras-related protein Rab-34; Ras-related protein Rab-34, isoform NARR; Ras-related protein Rab-39; ras-related protein Rah
Gene Symbols RAB34
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB34(Accession Q9BZG1), corresponding to amino acid residues K144-Y194.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360571, 83871
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.