Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB34 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735667 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5667 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5667 Supplier Proteintech Group Inc Supplier No. 274351AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein transport. Involved in the redistribution of lysosomes to the peri-Golgi region. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.

Specifications

Antigen RAB34
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB34
Gene Accession No. P0DI83
Gene Alias RAB34, RAB39, RAH, Ras related protein Rab 34, Ras related protein Rab 39, Ras related protein Rah
Gene Symbols RAB34
Host Species Rabbit
Immunogen RAB34 Fusion Protein Ag26696
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83871
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.