Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103244 Supplier Invitrogen™ Supplier No. PA5103244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB35.

RAB35 possesses GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB35
Gene Accession No. Q15286, Q5U316, Q6PHN9
Gene Alias 9530019H02Rik; AU040256; GTP-binding protein RAY; H-ray; I79_001225; RAB1C; Rab35; RAB35, member RAS oncogene family; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35; RAY
Gene Symbols RAB35
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB35(Accession Q15286), corresponding to amino acid residues N98-E148.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11021, 288700, 77407
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.